Hydroxylammonium sulphate ≥99.0%, J.T.Baker®

Danger

동의어: Hydroxylamine sulphateBis(hydroxylammonium) sulphateHydroxylamine sulphate (2:1)

Management of Change (MOC) category = N

N646-07EA 0 KRW
N646-07
Hydroxylammonium sulphate ≥99.0%, J.T.Baker®
Hydroxylammonium sulphate
포뮬러: H₈N₂O₆S
MW: 164,14 g/mol
녹는점: 170 °C
밀도: 1,1 g/cm³ (20 °C)
MDL 번호: MFCD00044869
CAS 번호: 10039-54-0
EINECS: 233-118-8
UN: 2865
ADR: 8,III
Merck 색인: 13,04853

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay ((NH₂OH)₂·H₂SO₄) ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR