trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%, Macron Fine Chemicals™

공급업체: Avantor Performance Materials
Warning

동의어: Titriplex® IV

MACR2357-57EA 0 KRW
MACR2357-57 MACR2357-55
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%, Macron Fine Chemicals™
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

For Laboratory, Research,or Manufacturing Use.

포뮬러: C₁₄H₂₂N₂O₈·1H₂O
MW: 364,35 g/mol
녹는점: 210 °C
보관 온도: ambient
MDL 번호: MFCD00066429
CAS 번호: 125572-95-4
EINECS: 236-308-9
UN: 0000

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay (C14H22N2O8 • H2O) ≥98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR