EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL), J.T.Baker®
공급업체: Avantor
동의어: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIEdetic acid disodium salt dihydrateKestranal® 2S dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate
Management of Change (MOC) category = NI
5648-07EA 750000 KRW
5648-07 5648-02 5648-03
EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL), J.T.Baker®
EDTA disodium salt solutions
Standardised at 25 °C against calcium carbonate whose certification is traceable to NIST.
포뮬러: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372,24 g/mol
MDL 번호: MFCD00150037
CAS 번호: 6381-92-6
Order Now
사양 테스트 결과
For Laboratory,Research,or Manufacturing Use
Titer, mg/mL, For Information Only
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Each lot of this product is standardized at 25°C against NIST StandardReference Material Calcium Carbonate (Hydroxy Naphthol Blue endpoint).
SRM No…
The titer found falls within the range 0.980-1.020 mg CaCO₃/mL.