Sodium hypophosphite monohydrate 99.0-105.0%, crystals, BAKER ANALYZED®, J.T.Baker®

동의어: Sodium phosphinate monohydrate

Management of Change (MOC) category = N

3741-01EA 532520 KRW
3741-01
Sodium hypophosphite monohydrate 99.0-105.0%, crystals, BAKER ANALYZED®, J.T.Baker®
Sodium hypophosphite monohydrate
포뮬러: NaH₂PO₂·H₂O
MW: 105,99 g/mol
녹는점: 90 °C
MDL 번호: MFCD06252379
CAS 번호: 10039-56-2
EINECS: 231-669-9
UN: 0000

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay (NaH₂PO₂·H₂O) 99.0 - 105.0 %
Insoluble Matter ≤ 0.01 %
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.04 %
Trace Impurities - Arsenic (As) ≤ 1 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR