Cobalt(II) sulphate heptahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent nickel-free

Danger

동의어: Cobalt sulphate heptahydrateCobaltous sulphate heptahydrate

22929.188EA 603510 KRW
22929.188
Cobalt(II) sulphate heptahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent nickel-free
Cobalt(II) sulphate heptahydrate
포뮬러: CoSO₄·7H₂O
MW: 281,1 g/mol
녹는점: 735 °C
밀도: 2,03 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00149658
CAS 번호: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck 색인: 12,02510

Order Now

사양 테스트 결과

Assay Min. 99.0 %
Cl (Chloride) Max. 10 ppm
Total N (Nitrogen) Max. 20 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 100 ppm
Ni (Nickel) Max. 50 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR