Diyttrium trioxide ≥99.9%, GPR RECTAPUR®

공급업체: VWR Chemicals

29033.101EA 105100 KRW
29033.101
Diyttrium trioxide ≥99.9%, GPR RECTAPUR®
Diyttrium trioxide
포뮬러: Y₂O₃
MW: 225.81 g/mol
끊는점: 4300 °C (1013 hPa)
녹는점: 2410 °C
밀도: 5,01 g/cm³ (20 °C)
MDL 번호: MFCD00011473
CAS 번호: 1314-36-9
EINECS: 215-233-5
UN: 0000

Order Now

사양 테스트 결과

Assay Min. 99.9 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR