Silver sulphate, BAKER ANALYZED® ACS, J.T. Baker®

공급업체: Avantor
Danger

동의어: Disilver sulphate

1183.0100EA 0 KRW
1183.0100 1183.0025
Silver sulphate, BAKER ANALYZED® ACS, J.T. Baker®
Silver sulphate
포뮬러: Ag₂SO₄
MW: 311.8 g/mol
끊는점: 1085 °C (1013 hPa)
녹는점: 655 °C
밀도: 5.45 g/cm³ (20°C)
보관 온도: Ambient
MDL 번호: MFCD00003407
CAS 번호: 10294-26-5
EINECS: 233-653-7
UN: 0000
Merck 색인: 12,08676
https://kr.vwr.com/stibo/web/std.lang.all/94/57/2299457.jpg

Order Now

사양 테스트 결과

Exceeds ACS Specifications
Assay min. 99.5 %
Iron (Fe) max. 0.001 %
Insoluble Matter and Silver Chloride max. 0.02 %
Nitrate (NO3) max. 0.001 %
Substances not Precipitated by HCl max. 0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR