Tannic acid, powder, BAKER ANALYZED®, J.T.Baker®

Danger

동의어: TanninGallotannin

0377-01EA 1011200 KRW
0377-01
Tannic acid, powder, BAKER ANALYZED®, J.T.Baker®
Tannic acid

  • Lot analysis on label

포뮬러: C₇₆H₅₂O₄₆
MW: 1701,2 g/mol
녹는점: 218…219 °C
밀도: 2,477 g/cm³ (20 °C)
인화점: 199 °C
보관 온도: Ambient
MDL 번호: MFCD00011386
CAS 번호: 1401-55-4
EINECS: 231-781-8
UN: 0000
ADR: 5.1,III
Merck 색인: 12,07825

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Residue after Ignition ≤0.10 %
Loss on Drying at 105°C ≤12.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR