Gentamicin sulphate 50 mg/ml
동의어: GaramycinGentiomycin CGentamicin sulphate salt
L0012-100EA 0 KRW
L0012-100 L0012-010
Gentamicin sulphate 50 mg/ml
Gentamicin sulphate
Gentamicin sulfate acts against gram-negative, gram-positive bacteria and mycoplasma.
  • Storage temperature of –20 °C
  • pH of 5±1
  • Shelf life of 24 months
  • Endotoxin: <10 EU/ml (<1 ng/ml)
Used in cell culture applications at 1 ml/l. This concentration is for tissue culture media containing serum; serum-free media generally require lower concentration.
주의: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
MW: 1530,9 g/mol
녹는점: 218 to 237 °C
보관 온도: ambient
MDL 번호: MFCD00270181
CAS 번호: 1405-41-0
EINECS: 215-778-9
Merck 색인: 13,04403