Calcium sulphate dihydrate ≥98%, GPR RECTAPUR®, precipitated

동의어: Plaster of Paris

22451.360EA 395850 KRW
22451.360
Calcium sulphate dihydrate ≥98%, GPR RECTAPUR®, precipitated
Calcium sulphate dihydrate
포뮬러: CaSO₄·2H₂O
MW: 172,17 g/mol
밀도: 2,32 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00010912
CAS 번호: 10101-41-4
EINECS: 231-900-3
UN: 0000
Merck 색인: 13,01711

Order Now

사양 테스트 결과

Assay (complexometric) Min. 98 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 100 ppm
Fe (Iron) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR