Dipotassium hydrogen orthophosphate trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
동의어: Potassium hydrogen phosphate trihydratePotassium phosphate dibasic trihydratedi-Potassium hydrogen orthophosphate trihydratedi-Potassium hydrogen phosphate trihydrate
103494GEA 267240 KRW
103494G 103495H
Dipotassium hydrogen orthophosphate trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Dipotassium hydrogen orthophosphate trihydrate
포뮬러: K₂HPO₄·3H₂O
MW: 228,22 g/mol
끊는점: < 340 °C (1013 hPa)
밀도: 2,504 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00149926
CAS 번호: 16788-57-1
EINECS: 231-834-5
UN: 0000
Order Now
사양 테스트 결과
Assay (acidimetric) Min. 99.0 %
pH (5 %) 9.2 - 9.4
Solution in water Passes test
Cl (Chloride) Max. 0.001 %
SO4 (Sulphate) Max. 0.005 %
Total N (Nitrogen) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
As (Arsenic) Max. 0.00005 %
Cu (Copper) Max. 0.0003 %
Fe (Iron) Max. 0.0005 %
Na (Sodium) Max. 0.1 %
Pb (Lead) Max. 0.001 %