Glutaraldehyde 50% in aqueous solution, Reagent Grade
|
Danger
|
M155-100MLEA
292200
KRW
M155-100ML
Glutaraldehyde 50% in aqueous solution, Reagent Grade
Glutaraldehyde
포뮬러:
C₅H₈O₂ MW: 100,12 g/mol 끊는점: 188 °C (1013 hPa) 녹는점: -10 °C 밀도: 1,13 g/cm³ (20 °C) 보관 온도: Refrigerator |
MDL 번호:
MFCD00007025 CAS 번호: 111-30-8 EINECS: 203-856-5 UN: 3265 ADR: 8 (6.1),II Merck 색인: 13,04485 |
사양 테스트 결과
Boiling Range | >95°C |
Color (APHA) | <100 |
Glutaraldehyde | 49 - 52% |
Methanol | <1% |
pH @25 °C | <5 |
Solidification Range | > –20 °C |
Specific Gravity | 1.125 - 1.135 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...