VWR®, Disposable Bacteriological Pipettes
공급업체: VWR
VWR®
VWRU53281-816EA 195270 KRW
VWRU53281-816 VWRU53281-818
VWR®, Disposable Bacteriological Pipettes
피펫 Bacteriological Pipettes
These sterile glass pipettes with wide tips are excellent for transferring viscous or suspended liquids.
  • Calibrated to deliver to the tip
  • Two rings near the top denote that the small amount remaining in the tip after free delivery must be blown out and added to the main volume delivered
  • Provided with negative graduations to increase the versatility of the pipette
  • Sterile, plugged
납품 정보: Supplied in multi-pack bags to protect against contamination.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR