VWR®, Mini Coolers, −20 °C
공급업체: VWR
VWR®
VWRU414004-286EA 234397 KRW
VWRU414004-286 VWRU414004-287 VWRU414004-285
VWR®, Mini Coolers, −20 °C
박스 Cooling/Freezing Boxes and Bags
These mini coolers are an alternate to ice buckets for keeping reagents and enzymes cool.
  • Coolers and lids are made of PC
  • Holds 0,5 or 1,5 ml microcentrifuge tubes
  • Protects critical samples from temperature fluctuations in the freezer
  • Cooler must be stored at –20 to –25 °C for 24 hours before use