VWR®, Sleeves for Ground Joints, PTFE
공급업체: VWR
VWR®
VWRU89026-480EA 40770 KRW
VWRU89026-480 VWRU89026-490 VWRU89026-488 VWRU89026-484
VWR®, Sleeves for Ground Joints, PTFE
조인트 슬리브 및 커넥터 Joint Sleeves
These PTFE sleeves withstand low to medium pressure and are inserted between standard ground glass joints to eliminate the use of grease and prevent sticking.
  • Chemically resistant
  • Inert
  • Usable upto 280 °C (536 °F)
Sleeves are inert, have a wall thickness of about 0,05 mm.