VWR®, Precision tweezers
공급업체: VWR
VWR®
VWRU89259-984EA 82230 KRW
VWRU89259-984
VWR®, Precision tweezers
집게
These precision tweezers are made from stainless steel and feature very sharp, fine points.
  • Anti-magnetic properties
  • Resistant to acids