D-(-)-Fructose, GenAR® for biotechnology, Macron Fine Chemicals™

동의어: D(-)-Levuloseβ-D-(-)-FructoseFructose

7756-12
D-(-)-Fructose, GenAR® for biotechnology, Macron Fine Chemicals™
D-(-)-Fructose
포뮬러: C₆H₁₂O₆
MW: 180,16 g/mol
끊는점: 552 °C (1013 hPa)
녹는점: 100 to 110 °C
보관 온도: ambient
MDL 번호: MFCD00148910
CAS 번호: 57-48-7
EINECS: 200-333-3
UN: 0000
Merck 색인: 13,04295
https://kr.vwr.com/stibo/web/std.lang.all/96/14/2299614.jpg

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₂O₆) 98.0 - 102.0 %
Acidity Passes Test
Arsenic (As) ≤0.0001 %
Calcium and Magnesium ≤0.005 %
Chloride (Cl) ≤0.018 %
Color of Solution Passes Test
Glucose ≤0.2 %
Heavy Metals (as Pb) ≤0.0005 %
Identification Passes Test
Loss on Drying ≤0.5 %
Residue on Ignition ≤0.5 %
Sulfate (SO₄) ≤0.025 %
Hydroxymethylfurfural Passes Test
Appearance (colorless crystals or white crystalline powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR