J.T.Baker® BAKER INSTRA-ANALYZED®, Boron Standard Solution, 1000 µg/L B in Water, Reagent for AAS
공급업체: Avantor
6446-04EA 0 KRW
BAKR6446-04
J.T.Baker® BAKER INSTRA-ANALYZED®, Boron Standard Solution, 1000 µg/L B in Water, Reagent for AAS
Boron standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Prepared from H₃BO₃H
증명: Traceable to NIST and SRM numbers.