Dry shippers, DS series
VWRI478-0305EA 1207870 KRW
VWRI478-0305 VWRI478-0317 VWRI478-0318 VWRI478-0319
Dry shippers, DS series
Dry shippers are used for the safe transport of samples at cryogenic temperatures.
Hydrophobic absorbent material is fixed in the inner capacity of the vessel. After complete saturation of the material with liquid nitrogen, the vessels are ready to transport samples at cryogenic temperatures, without the risk of liquid nitrogen spilling if the container is overturned.
Model DS-24 can be equipped with removable absorbent cartridges to improve the holding time.
* DS-24 is delivered without canister, rack or cartridge (must be ordered separately). For details of the range contact your local VWR sales office.
증명: CE - Medical.
Shippers do not conform to International Air Transport Regulations (IATA) for transportation of infectious material.