Standards for the determination of Hydrocarbon Oil Index (HOI)
VWRC87498.005EA 381710 KRW
VWRC87498.005
Standards for the determination of Hydrocarbon Oil Index (HOI)
Standards designed for the analysis of water and ground samples.
증명: ISO 9377-2.
납품 정보: Delivered with Certificate of Analysis, reference and batch number of source materials.
포장: 1,0 and 10 ml glass ampoules.