VWR®, Deep Well Microplates, 96-Well
공급업체: VWR
VWR®
VWRU89237-526EA 87570 KRW
VWRU89237-526 VWRU89237-514 VWRU89237-516 VWRU89237-522 VWRU89237-518
VWR®, Deep Well Microplates, 96-Well
마이크로플레이트 Deep Well Microplates
PP. These microplates are uniform dimensionally so that all covers, whether round or square, fit interchangeably on other round or square well microplates. This reduces the number of covers needed for most laboratories. The deep well microplates are available in round well with round bottoms for volumes up to 1,0 ml and square well with either a conical or round bottom for volumes up to 2,0 ml. Also available in shallow well and medium well formats.