Buffered oxide etch (5:1), CMOS for microelectronic use, J.T.Baker®
|
Danger
|
5192-03EA
0
KRW
5192-03
Buffered oxide etch (5:1), CMOS for microelectronic use, J.T.Baker®
Buffered oxide etch
UN:
2922 ADR: 8,II |
사양 테스트 결과
For Microelectronic Use | |
Ship and store material above 70°F (21°C) | |
Ammonium Fluoride (NH₄F)(w/w) | 32.0 - 34.0 % |
Hydrofluoric Acid (HF)(w/w) | 8.2 - 8.5 % |
Residue after Ignition | ≤10 ppm |
Chloride (Cl) | ≤3000.0 ppb |
Nitrate (NO₃) | ≤3000 ppb |
Phosphate (PO₄) | ≤700.0 ppb |
Sulfate (SO₄) | ≤1500.0 ppb |
Trace Impurities - Aluminum (Al) | ≤50.0 ppb |
Arsenic and Antimony (as As) | ≤30.0 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤20.0 ppb |
Trace Impurities - Boron (B) | ≤50 ppb |
Trace Impurities - Cadmium (Cd) | ≤20.0 ppb |
Trace Impurities - Calcium (Ca) | ≤100.0 ppb |
Trace Impurities - Chromium (Cr) | ≤10.0 ppb |
Trace Impurities - Cobalt (Co) | ≤10.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤40.0 ppb |
Trace Impurities - Gold (Au) | ≤20 ppb |
Heavy Metals (as Pb) | ≤1000.0 ppb |
Trace Impurities - Iron (Fe) | ≤50.0 ppb |
Trace Impurities - Lead (Pb) | ≤10.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤50.0 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Molybdenum (Mo) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤10.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Silver (Ag) | ≤50 ppb |
Trace Impurities - Sodium (Na) | ≤100.0 ppb |
Trace Impurities - Strontium (Sr) | ≤10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤20.0 ppb |
Trace Impurities - Tin (Sn) | ≤20.0 ppb |
Trace Impurities - Titanium (Ti) | ≤100.0 ppb |
Trace Impurities - Zinc (Zn) | ≤100.0 ppb |
Particle Count - 0.5 µm and greater | ≤150 par/ml |
Particle Count - 1.0 µm and greater | ≤8 par/ml |
For additional information, go to www.askavantor.com. Search keywords | |
freezing" and product name." |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...