Ethanol denatured (denatured with 4% MIBK and kerosene), Macron Fine Chemicals™
|
Danger
|
MACR7018-19EA
0
KRW
MACR7018-19
7018-16
Ethanol denatured (denatured with 4% MIBK and kerosene), Macron Fine Chemicals™
Ethanol denatured
Made from Completely Denatured Alcohol Formula 19
- Total alcohol and denaturant: 95%
- Ethyl alcohol: 90%
- Denaturant: 4% methyl isobutyl ketone and \<1% kerosene
포뮬러:
H₃CCH₂OH MW: 46,07 g/mol 보관 온도: Ambient |
MDL 번호:
MFCD00003568 CAS 번호: 64-17-5 ADR: 3,II |
사양 테스트 결과
For Laboratory,Research,or Manufacturing Use | |
Identification (by GLC) | Passes Test |
Residue after Evaporation (w/v) | ≤0.003 % |
CDA Formula 19 consists of 4 volumes of methyl isobutyl ketone, 1 | |
volumeof kerosene, and 100 volumes of 190 proof ethanol. | |
Product Information (not specifications): Appearance (clear, colorless | |
liquid) |