Eriochrome Black T, powder, BAKER ANALYZED® ACS, J.T.Baker®

Warning

동의어: Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonateEBTSolochrome Black T

EXPOJTL126-3 L126-03
Eriochrome Black T, powder, BAKER ANALYZED® ACS, J.T.Baker®
Eriochrome Black T

Powder.

포뮬러: C₂₀H₁₂N₃NaO₇S
MW: 461,39 g/mol
보관 온도: Ambient
MDL 번호: MFCD00003935
CAS 번호: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
Merck 색인: 12,03709

Order Now

사양 테스트 결과

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Clarity of Solution Passes Test
ACS - Suitability as a Complexometric Indicator Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR