Sodium nitroprusside dihydrate 99.0-102.0%, BAKER ANALYZED® ACS, J.T.Baker®

Danger

동의어: Sodium nitroferricyanide(III) dihydrateSodium nitrosopentacyanoferrate (III) dihydrateSodium nitroferricyanide dihydrateSodium pentacyanonitrosylferrate(III) dihydrateDisodium pentacyanonitrosylferrate (III) dihydrate

3792-04EA 0 KRW
3792-04
Sodium nitroprusside dihydrate 99.0-102.0%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium nitroprusside dihydrate

Lot analysis on label.

포뮬러: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297,95 g/mol
밀도: 1,72 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00149192
CAS 번호: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck 색인: 12,08794

Order Now

사양 테스트 결과

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂Fe(CN)₅NO·2H₂O) 99.0 - 102.0 %
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤0.02 %
Sulfate (SO₄) Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR