Dipotassium hydrogen orthophosphate trihydrate, crystals, AR®, Macron Fine Chemicals™
동의어: Potassium hydrogen phosphate trihydratePotassium phosphate dibasic trihydratedi-Potassium hydrogen orthophosphate trihydratedi-Potassium hydrogen phosphate trihydrate
MACR7088-06EA 0 KRW
MACR7088-06 7088-04
Dipotassium hydrogen orthophosphate trihydrate, crystals, AR®, Macron Fine Chemicals™
Dipotassium hydrogen orthophosphate trihydrate
포뮬러: K₂HPO₄·3H₂O
MW: 228,22 g/mol
끊는점: ∼340 °C (1013 hPa)
밀도: 2,504 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00149926
CAS 번호: 16788-57-1
EINECS: 231-834-5
UN: 0000
Order Now
사양 테스트 결과
For Laboratory,Research,or Manufacturing Use
Solubility (10 in 100) Passes Test
Chloride (Cl) ≤0.003 %
Heavy Metals (as Pb) ≤0.0005 %
Insoluble Matter ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Nitrogen (N) ≤0.001 %
Sodium (Na) ≤0.02 %
Sulfate (SO₄) ≤0.005 %
pH of 0.1M Solution at 25°C 9.0 - 9.4
Water (H₂O), For Information Only
Appearance (colorless crystals)
Learn more