Chloramine-T (sodium salt) trihydrate, J.T.Baker®

공급업체: Avantor
Danger

동의어: Chloramine T sodium salt trihydrateTosylchloramide sodium trihydrate

E494-06EA 1148000 KRW
E494-06
Chloramine-T (sodium salt) trihydrate, J.T.Baker®
Chloramine-T (sodium salt) trihydrate
포뮬러: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281,69 g/mol
녹는점: 167…170 °C
밀도: 1,43 g/cm³ (20 °C)
인화점: 192 °C (closed cup)
보관 온도: Ambient
MDL 번호: MFCD00149066
CAS 번호: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8,III
Merck 색인: 13,02085

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Active Chlorine ≥11 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR