4-(Methylamino)phenol sulphate 99.0-101.5%, J.T.Baker®

Warning

동의어: Metol

Q067-07EA 0 KRW
Q067-07
4-(Methylamino)phenol sulphate 99.0-101.5%, J.T.Baker®
4-(Methylamino)phenol sulphate

Lot analysis on label.

포뮬러: C₁₄H₂₀N₂O₆S
MW: 344,39 g/mol
끊는점: 227 °C (1013 hPa)
녹는점: 86 °C
밀도: 1,434 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00013140
CAS 번호: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck 색인: 12,06097

Order Now

사양 테스트 결과

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃NHC₆H₄OH)₂H₂SO₄) 99.0 - 101.5 %
Residue after Ignition ≤ 0.10 %
Suitability for Phosphate Determination Passes Test
Solubility in HCl Passes Test
o-Aminophenol Passes Test
Chloride (Cl) Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR