Sulphanilamide 90-100%, J.T.Baker®

Warning

동의어: 4-Aminobenzenesulphonamide

V153-05EA 453280 KRW
V153-05
Sulphanilamide 90-100%, J.T.Baker®
Sulphanilamide
포뮬러: H₂NC₆H₄SO₂NH₂
MW: 172,21 g/mol
끊는점: 401 °C (1013 hPa)
녹는점: 163…167 °C
밀도: 1,46 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00007939
CAS 번호: 63-74-1
EINECS: 200-563-4
UN: 0000
Merck 색인: 12,09094
https://kr.vwr.com/stibo/web/std.lang.all/76/03/2297603.jpg

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Melting Point 163 - 166 °C
Not intended for food, animal, food chain or feed applications.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR