VWR®, Spatulas, Stainless Steel/PVC Handle
공급업체: VWR
VWR®
VWRU82027-502EA 407070 KRW
VWRU82027-502 VWRU82027-506 VWRU82027-516
VWR®, Spatulas, Stainless Steel/PVC Handle
주걱
Stainless steel, PVC handle. Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • PVC handle is resistant to acids and most solvents
  • Handle can withstand boiling water
  • Multiple sizes allow for varied laboratory applications