VWR®, Benchtop chillers
공급업체: VWR
VWR®
VWRU89004-560EA 493470 KRW
VWRU89004-560
VWR®, Benchtop chillers
쿨러
냉각장치
Benchtop Coolers
Polycarbonate and thermal gel chillers designed to protect frozen samples by helping to maintain freezer temperatures on the lab bench.