VWR®, Absorbent Pads
공급업체: VWR
VWR®
VWRU95057-860EA 299320 KRW
VWRU95057-860
VWR®, Absorbent Pads
유출 통제 물질
Designed to soak up liquids, oils, and common solvents, absorbent pads provide an excellent alternative to paper towels and gauze.
  • Strong, highly absorbent, and lint-free
  • Efficient alternative to paper towels and gauze
Pads exhibit a cloth-like softness with no loose threads or fibers, making them ideal for use in coverslipping, slide preparation, and general laboratory cleanup.