VWR® Talon®, Suspension clamp
공급업체: VWR
VWRU21573-153EA 840450 KRW
VWRU21573-153
VWR® Talon®, Suspension clamp
클램프 General Purpose Clamps
Clamp holds thermometers, potash bulbs or drying tubes 114 mm from the support rod. Machine tapered, nickel plated brass hook won't bend, rust or corrode. Built in holder grips rods up to 19 mm in Ø.