VWR®, Bench Protectors with Leakproof Moisture Barrier
공급업체: VWR
VWR®
VWRU89126-794EA 453207 KRW
VWRU89126-794 VWRU89126-790 VWRU89126-792
VWR®, Bench Protectors with Leakproof Moisture Barrier
표면 보호 용지 Bench Protectors
These super absorbent bench protectors feature a two-layer, cushioned construction for use in a laboratory or veterinary setting. The air-lay cellulose layer provides cushion for laboratory equipment and glassware to prevent breakage, while the leakproof plastic backsheet protects tables and benches from spills and moisture.
  • Two layers of protection and leakproof backsheet
  • Available as rolls or mats
  • Can be cut to desired size