VWR®, Double Spatulas, Hayman Style
공급업체: VWR
VWR®
VWRU82027-524EA 412540 KRW
VWRU82027-524 VWRU82027-522 VWRU82027-522_U
VWR®, Double Spatulas, Hayman Style
주걱
Stainless steel. These spatulas are ideal for applications involving the transfer or mixing of small sample amounts.
Model 82027-522 features a round spatula on one end and a spoon on the other. Model 82027-524 features a tapered, bent-tip spatula on one end and a spoon on the other. Efficient design reduces the risk of sample loss.