VWR®, Dishes, Crystallising
공급업체: VWR
VWR®
VWRU10754-770EA 247860 KRW
VWRU89000-286 VWRU89000-296 VWRU10754-770 VWRU89000-294 VWRU10754-782 VWRU89000-280
VWR®, Dishes, Crystallising
트레이/디쉬/포트 Crystallising Dishes
Borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for storage and crystallisation. Rims reinforced and fire-polished to reduce chipping.