VWR®TraceClean®, Water sampling bottles, glass, wide neck
공급업체: VWR
VWR® TraceClean®
VWRU89093-958EA 102760 KRW
VWRU89093-958 VWRU89094-030 VWRU89093-998 VWRU89093-976 VWRU89093-978 VWRU89093-956 VWRU89094-012 VWRU89094-010 VWRU89094-006 VWRU89094-014 VWRU89094-008 VWRU89093-950 VWRU89093-952 VWRU89093-980 VWRU89093-960 VWRU89093-982
VWR®TraceClean®, Water sampling bottles, glass, wide neck
바틀 Water Sampling Bottles
Type III (soda-lime) glass.
  • Available with solid-top or open-top caps
  • Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, PCBs, and phenolics analyses
  • Open-top caps with ultrasonically bonded fluoropolymer resin/silicone septa are for use in volatile organic analysis
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.