VWR®, Biohazard Labels
공급업체: VWR
VWR®
VWRU89097-876EA 63520 KRW
VWRU89097-876 VWRU89097-852
VWR®, Biohazard Labels
라벨
Labels aid in general workplace safety and efficiency by quickly and simply identifying contaminated equipment containers, rooms, or materials. A biohazard symbol along with the term are printed in bold black atop a fluorescent red background.
  • Manufactured in accordance with various federal and state regulations
  • Including OSHA safety standards
주문 정보:Labels are available in rolls of 250 or 500 labels. One roll per case.