VWR®, Crucible Tongs, Stainless Steel
공급업체: VWR
VWRU82027-362EA 1000930 KRW
VWRU82027-362 VWRU82027-368
VWR®, Crucible Tongs, Stainless Steel
집게
Stainless steel. These tongs are designed for handling a wide variety of laboratory apparatus.
Model 82027-362 is a general purpose tong with serrated tips. Model 82027-368 features a nickel-plated steel construction and an extra long design.