VWR®, Dishes, Evaporating, with Spout
공급업체: VWR
VWR®
VWRU89038-082EA 149733 KRW
VWRU89038-082 VWRU89038-092 VWRU89038-108 VWRU89038-104 VWRU89038-078 VWRU89038-088 VWRU89038-086
VWR®, Dishes, Evaporating, with Spout
트레이/디쉬/포트 Evaporating Basins/Dishes
Porcelain, glased inside and out, except base and rim.
  • Resistant to chemicals
  • Withstand maximum temperature of 1150 °C