VWR®, Gooch Crucibles
공급업체: VWR
VWR®
VWRU89038-038EA 273070 KRW
VWRU89038-038 VWRU89038-046 VWRU89038-042
VWR®, Gooch Crucibles
도자기 용기
Porcelain gooch crucibles, with perforated bottom, glazed inside and out, except base and rim. Ideal for total suspended solids determination. Crucible 89038-046 has two holes for suspending in extraction apparatus.
  • For use in temperatures up to 1150 °C
주문 정보:Lids need to be ordered seperately.