VWR® Traceable® One-Shot™, Conductivity standards (CRM)
공급업체: Avantor
Traceable® VWR® Traceable® One-Shot™
VWRU23226-567EA 246592 KRW
VWRU23226-653 VWRU23226-654 VWRU23226-567 VWRU46610-042 VWRU46610-032 VWRU12777-828 VWRU23226-650 VWRU23226-651 VWRU23226-652 VWRU12777-832 VWRU46610-040 VWRU46610-030 VWRU36934-134 VWRU89030-248 VWRU62344-936 VWRU36934-136
VWR® Traceable® One-Shot™, Conductivity standards (CRM)
표준 물질 Conductivity Standards
Traceable® Conductivity Standards are among the most accurate available. They are compatible with all makes of equipment, and can be used in lab or field. Each bottle is labeled for calibrating conductivity (microsiemens/micromhos), resistivity (ohms), and dissolved solids (parts per million).
  • Certified reference material
  • 100% compatible with all makes of instruments and probes
  • Calibration within disposable container eliminates contamination
증명: Accredited ISO Guide 34, ISO Guide 35, ISO Guide 31, ISO/IEC 17025.
납품 정보: Supplied with 480 ml container, step-by-step calibration instructions, individual temperature compensation charts, traceability information and Traceable® certificate.