VWR®, Aluminum Foil, Premium, Ultra-Clean
공급업체: VWR
VWR® UltraClean™
VWRU89107-730EA 116090 KRW
VWRU89107-730 VWRU89107-732 VWRU89107-734 VWRU89107-724 VWRU89107-736 VWRU89107-726
VWR®, Aluminum Foil, Premium, Ultra-Clean
알루미늄 호일
Premium, ultra-clean aluminium foil is strong and puncture-resistant.
  • Available in a variety of formats
  • Contains 200 sheets
  • Puncture-resistant
Foil is able to withstand extreme heat and cold, and has the lowest moisture-vapour transfer rate of all wrapping materials. Standard, heavy-duty, and extra heavy-duty formats are supplied as boxed rolls with a metal tear-off bar for ease in obtaining exact size requirements. The pop-up dispenser contains 200 individual sheets.
납품 정보: Supplied as boxed rolls with a metal tear-off bar for ease in obtaining exact size requirements.