VWR®, Connectors, Tubing, Tapered
공급업체: VWR
VWR®
VWRU82017-256EA 261490 KRW
VWRU82017-256 VWRU46600-118 VWRU46600-116 VWRU46600-078 VWRU46600-076 VWRU46600-096 VWRU46600-074
VWR®, Connectors, Tubing, Tapered
이음쇠 부속품
PP, white.
  • Excellent chemical and temperature resistance
  • Will withstand flow up to 1,37 bar
  • Autoclavable
The arms of these connectors are tapered to accommodate a wide variety of tubing sizes.