VWR® VistaVision, Cover Glasses
공급업체: VWR
VWR® VistaVision™ VWR VistaVision™ VistaVision™
VWRU16004-348EA 167730 KRW
VWRU16004-348 VWRU16004-314 VWRU16004-306 VWRU16004-346 VWRU16004-312 VWRU16004-340 VWRU16004-098 VWRU16004-318 VWRU16004-096
VWR® VistaVision, Cover Glasses
커버 글라스
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.
  • No disturbing self-fluorescence under UV light
  • Suitable for use in coverslippers
증명: ISO 8255-1
납품 정보: Rectangular cover glasses No. 1 are supplied 100 per hinged box. Rectangular cover glasses No. 1.5 are supplied 1 oz. per hinged box.