Avantor® ACE® Excel® Silica, HPLC/UHPLC Columns, Analytical, 3 µm
공급업체: VWR
Avantor® ACE® Excel®
EXL-117-2503UEA 1528880 KRW
EXL-117-2503U EXL-117-1503U EXL-117-2546U
Avantor® ACE® Excel® Silica, HPLC/UHPLC Columns, Analytical, 3 µm
크로마토그래피 컬럼
Avantor® ACE® Excel® Silica columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Wide range of particle sizes (2 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
For additional dimensions please also see the Avantor® ACE® Silica, 3 μm product page.