Resin, medical grade.
- Tear-open film package designed for use with gloves
- Validated secure medical-grade seal for purity
- Certified free from endotoxin, RNase, DNase and ATP
- Sustainable design based on virgin resins
VWR 15 and 50 ml sterile centrifuge tubes are packed in sleeves of 10, bringing a new level of convenience and purity in your centrifuge labware. Each pack of 50 tubes is neatly wrapped and compact. Comprised of 5 sections of 10 tubes, each of the subpacks can be separated without scissors and used anywhere in the lab. Because these sleeves are loaded robotically, each tube is perfectly ordered unlike competing brands. Every subpack is labelled with a lot number and sterility expiration date. Each pack has a tear point at the seam, designed specifically for use with gloves. These sleeves of 10 feature a validated, secure, medical-grade seal that is so precise, it can withstand immersion without leakage. These subpacks are more convenient for cleanroom wash-down than competing packaging.