VWR®, Sterile Sample Bags
공급업체: VWR
VWR®
VWRU89085-538EA 433660 KRW
VWRU82007-706 VWRU83007-558 VWRU89085-538 VWRU89000-190 VWRU83007-556 VWRU82007-698 VWRU89085-528 VWRU82007-714 VWRU89085-556 VWRU82007-726 VWRU89085-558 VWRU89085-546 VWRU89085-530 VWRU89085-532 VWRU89085-554 VWRU89085-542 VWRU89085-580 VWRU83007-562 89004-420 VWRU89000-642 VWRU89000-640
VWR®, Sterile Sample Bags
Sample Bags
Bags made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals
  • The closure provides the strongest, most secure seal
  • With white marking areas allow written notation
These sample bags are ideal for transportation and storage of solids, semi-solids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
They feature a wire closure for an airtight seal, and wide-sealed bottoms for added leakproof protection. To use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
증명: Bags comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
주의: Note: Do not use with alcoholic substances or where temperature exceeds 100 °C (212 °F).
*The product(s) on this page are outside the scope of the EU Directive 2019/904