VWR®, Disposable Weighing Dishes, Aluminium, Smooth Sides
공급업체: VWR
25433-030EA 68853 KRW
25433-030 25433-089
VWR®, Disposable Weighing Dishes, Aluminium, Smooth Sides
시료 접시 Weighing Boats
Smooth walled, flat bottomed, aluminium dishes are ideal for general purpose weighing, dispensing, storing, sampling, and testing applications.
  • Oil-free and non-coated
  • No tab
  • Contoured to facilitate stacking and dispensing