Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®
공급업체: Avantor
동의어: Molecular sieve A3
Management of Change (MOC) category = N
2710-01EA 836000 KRW
2710-01 2710-05
Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®
Molecular sieve 3A (0.3 nm, 3 Å)
포뮬러: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL 번호: MFCD00131613
CAS 번호: 308080-99-1
EINECS: 000-000-0
UN: 0000
Order Now
사양 테스트 결과
For Laboratory,Research,or Manufacturing Use
Loss on Drying at 150°C ≤ 5 %